Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester, 97%

$71.12 - $240.12

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
CAS 1025718-78-8
Molecular Formula C14H18BBrO4
Formula Weight 341.01
MDL Number MFCD12405334
Synonym Methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51718MD

 
alfa aesar™
H51718MD
250mg Each for $71.12

AAH5171803

 
alfa aesar™
H5171803
1g Each for $240.12
Description & Specifications

Specifications

Formula Weight 341.01
Chemical Name or Material 3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C14H18BBrO4
MDL Number MFCD12405334
CAS 1025718-78-8
Melting Point 98° to 102°C
Synonym Methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Quantity 250mg