missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-iodopyridine, 95%

Manufacturer:  Alfa Aesar™ H64007MD

 View more versions of this product

Catalog No. AAH64007MD



Description

Description

Applications
Reactant in the synthesis chiral 4, 4′-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat, air.
Specifications

Specifications

3-Bromo-5-iodopyridine
White
250mg
C5H3BrIN
108478
AOOZLVWDZUPEHT-UHFFFAOYSA-N
3-bromo-5-iodopyridine
820409
Solid
Light Sensitive
127°C to 131°C
233770-01-9
MFCD03086019
3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
C1=C(C=NC=C1I)Br
283.894
283.89
95%

Chemical Identifiers

233770-01-9
283.894
AOOZLVWDZUPEHT-UHFFFAOYSA-N
820409
C1=C(C=NC=C1I)Br
C5H3BrIN
MFCD03086019
3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
3-bromo-5-iodopyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents