missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-5-(hydroxymethyl)pyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 37669-64-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
InChI Key WDVDHJLKXYCOFS-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxymethyl pyridine, 3-bromo-5-hydroxymethylpyridine, 3-bromo-pyridin-5-yl methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinemethanol, 5-bromo-3-pyridinyl methanol, 5-bromo-3-pyridyl methan-1-ol, 5-bromo-pyridin-3-yl-methanol, 5-bromopyridin-3-yl methanol
PubChem CID 2784733
IUPAC Name (5-bromopyridin-3-yl)methanol
SMILES C1=C(C=NC=C1Br)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444050010
SDS View Documents
Thermo Fisher Scientific
AC444050010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444050050
View Documents
Thermo Fisher Scientific
AC444050050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

37669-64-0
188.02
3-bromo-5-hydroxymethyl pyridine, 3-bromo-5-hydroxymethylpyridine, 3-bromo-pyridin-5-yl methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinemethanol, 5-bromo-3-pyridinyl methanol, 5-bromo-3-pyridyl methan-1-ol, 5-bromo-pyridin-3-yl-methanol, 5-bromopyridin-3-yl methanol
(5-bromopyridin-3-yl)methanol
C6H6BrNO
WDVDHJLKXYCOFS-UHFFFAOYSA-N
2784733
C1=C(C=NC=C1Br)CO

Specifications

Authentic
154.0°C (4.0 Torr)
27.0°C
37669-64-0
100.0
C6H6BrNO
WDVDHJLKXYCOFS-UHFFFAOYSA-N
(5-bromopyridin-3-yl)methanol
2784733
Liquid or Low Melting Solid
3-Bromo-5-(hydroxymethyl)pyridine
Glass Bottle
Yellow
1g
96.0
96% min. (GC)
3-bromo-5-hydroxymethyl pyridine, 3-bromo-5-hydroxymethylpyridine, 3-bromo-pyridin-5-yl methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinemethanol, 5-bromo-3-pyridinyl methanol, 5-bromo-3-pyridyl methan-1-ol, 5-bromo-pyridin-3-yl-methanol, 5-bromopyridin-3-yl methanol
C1=C(C=NC=C1Br)CO
188.02
188.02
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS