missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorophenylacetonitrile, 96%

Manufacturer:  Alfa Aesar™ H6260503

 View more versions of this product

Catalog No. AAH6260503



Specifications

Specifications

3-Bromo-5-fluorophenylacetonitrile
305800-58-2
MFCD09258960
2-3-bromo-5-fluorophenyl acetonitrile
C1=C(C=C(C=C1F)Br)CC#N
214.037
214.03
96%
1g
C8H5BrFN
UN3439
FSKMLBLIAOVQCF-UHFFFAOYSA-N
2-(3-bromo-5-fluorophenyl)acetonitrile
23509635
Solid

Chemical Identifiers

305800-58-2
214.037
FSKMLBLIAOVQCF-UHFFFAOYSA-N
23509635
C1=C(C=C(C=C1F)Br)CC#N
C8H5BrFN
MFCD09258960
2-3-bromo-5-fluorophenyl acetonitrile
2-(3-bromo-5-fluorophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents