missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorophenol, 97%

Manufacturer:  Alfa Aesar™ H5528803

 View more versions of this product

Catalog No. AAH5528803



Specifications

Specifications

3-Bromo-5-fluorophenol
36°C to 40°C
433939-27-6
MFCD07783710
JCPJGUPQZDEZQH-UHFFFAOYSA-N
3-bromo-5-fluorophenol
21904636
97%
>110°C (230°F)
1g
C6H4BrFO
3-bromo-5-fluoro-phenol
C1=C(C=C(C=C1F)Br)O
190.999
191

Chemical Identifiers

433939-27-6
190.999
JCPJGUPQZDEZQH-UHFFFAOYSA-N
21904636
C1=C(C=C(C=C1F)Br)O
C6H4BrFO
MFCD07783710
3-bromo-5-fluoro-phenol
3-bromo-5-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents