Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzyl bromide, 95%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-fluorobenzyl bromide
CAS 216755-57-6
Molecular Formula C7H5Br2F
Formula Weight 267.92
MDL Number MFCD09258959
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6376903

  SDS
alfa aesar™
H6376903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6376906

  SDS
alfa aesar™
H6376906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.92
Chemical Name or Material 3-Bromo-5-fluorobenzyl bromide
Assay 95%
Quantity 1g
Molecular Formula C7H5Br2F
MDL Number MFCD09258959
CAS 216755-57-6
UN Number UN3261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water (2.9 g/L) (25°C).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C