missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzyl bromide, 95%

Manufacturer:  Alfa Aesar™ H6376906

 View more versions of this product

Catalog No. AAH6376906



Description

Description

Applications
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water (2.9 g/L) (25°C).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

3-Bromo-5-fluorobenzyl bromide
216755-57-6
MFCD09258959
3-bromo-5-fluorobenzyl bromide, 1-bromo-3-bromomethyl-5-fluorobenzene, 3-fluoro-5-bromobenzyl bromide, 5-bromo-3-fluorobenzyl bromide, 3-fluoro-5-bromobenzylbromide, 5-bromo-3-bromomethyl-1-fluorobenzene, pubchem4886, acmc-20e3gr, 3-bromo-5-fluorobenzylbromide, benzene,1-bromo-3-bromomethyl-5-fluoro
C1=C(C=C(C=C1F)Br)CBr
267.923
267.92
5g
C7H5Br2F
UN3261
DAUWIPUGOIFZNF-UHFFFAOYSA-N
1-bromo-3-(bromomethyl)-5-fluorobenzene
23509619
95%

Chemical Identifiers

216755-57-6
267.923
DAUWIPUGOIFZNF-UHFFFAOYSA-N
23509619
C1=C(C=C(C=C1F)Br)CBr
C7H5Br2F
MFCD09258959
3-bromo-5-fluorobenzyl bromide, 1-bromo-3-bromomethyl-5-fluorobenzene, 3-fluoro-5-bromobenzyl bromide, 5-bromo-3-fluorobenzyl bromide, 3-fluoro-5-bromobenzylbromide, 5-bromo-3-bromomethyl-1-fluorobenzene, pubchem4886, acmc-20e3gr, 3-bromo-5-fluorobenzylbromide, benzene,1-bromo-3-bromomethyl-5-fluoro
1-bromo-3-(bromomethyl)-5-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

SDS Documents