missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ H6141506

 View more versions of this product

Catalog No. AAH6141506



Specifications

Specifications

3-Bromo-5-fluorobenzyl alcohol
5g
C7H6BrFO
3-bromo-5-fluoro-phenyl methanol
C1=C(C=C(C=C1F)Br)CO
205.026
205.02
98%
Colorless
216755-56-5
MFCD04116320
KQWBQTOZXPPFIY-UHFFFAOYSA-N
(3-bromo-5-fluorophenyl)methanol
2769402
Liquid

Chemical Identifiers

216755-56-5
205.026
KQWBQTOZXPPFIY-UHFFFAOYSA-N
2769402
C1=C(C=C(C=C1F)Br)CO
C7H6BrFO
MFCD04116320
3-bromo-5-fluoro-phenyl methanol
(3-bromo-5-fluorophenyl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents