missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-fluorobenzotrifluoride 98.0+%, TCI America™
$28.21 - $109.49
Chemical Identifiers
CAS | 130723-13-6 |
---|---|
Molecular Formula | C7H3BrF4 |
Molecular Weight (g/mol) | 242.999 |
MDL Number | MFCD00042498 |
InChI Key | LIGBGEJPUQBLTG-UHFFFAOYSA-N |
Synonym | 3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl |
PubChem CID | 2736323 |
IUPAC Name | 1-bromo-3-fluoro-5-(trifluoromethyl)benzene |
SMILES | C1=C(C=C(C=C1F)Br)C(F)(F)F |
Chemical Identifiers
130723-13-6 | |
242.999 | |
LIGBGEJPUQBLTG-UHFFFAOYSA-N | |
2736323 | |
C1=C(C=C(C=C1F)Br)C(F)(F)F |
C7H3BrF4 | |
MFCD00042498 | |
3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl | |
1-bromo-3-fluoro-5-(trifluoromethyl)benzene |
Specifications
130723-13-6 | |
139°C | |
MFCD00042498 | |
3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl | |
C1=C(C=C(C=C1F)Br)C(F)(F)F | |
242.999 | |
243.00 | |
Liquid |
Yellow | |
C7H3BrF4 | |
5 g | |
LIGBGEJPUQBLTG-UHFFFAOYSA-N | |
1-bromo-3-fluoro-5-(trifluoromethyl)benzene | |
2736323 | |
≥98.0% (GC) | |
3-Bromo-5-fluorobenzotrifluoride |
Safety and Handling
TSCA : No