missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-5-fluorobenzotrifluoride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435500050

 View more versions of this product

Catalog No. AC435500050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Bromo-5-fluorobenzotrifluoride
>110°C
1.51
Colorless to Yellow
130723-13-6
100.0
C7H3BrF4
1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluoro-trifluoromethylbenzene, 3-bromo-5-fluorobenzotrifluoride, 3-bromo-5-fluorotrifluoromethylbenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-fluoro-5-trifluoromethyl bromobenzene, 3-fluoro-5-trifluoromethylbromobenzene, 5-fluoro-3-bromobenzotrifluoride, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
C1=C(C=C(C=C1F)Br)C(F)(F)F
243
243
97%
1.5100g/mL
Authentic
146°C
5g
96.0
96% min. (GC)
MFCD00042498
LIGBGEJPUQBLTG-UHFFFAOYSA-N
1-bromo-3-fluoro-5-(trifluoromethyl)benzene
2736323
Liquid or Low Melting Solid

Chemical Identifiers

130723-13-6
243
LIGBGEJPUQBLTG-UHFFFAOYSA-N
2736323
C1=C(C=C(C=C1F)Br)C(F)(F)F
C7H3BrF4
MFCD00042498
1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluoro-trifluoromethylbenzene, 3-bromo-5-fluorobenzotrifluoride, 3-bromo-5-fluorotrifluoromethylbenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-fluoro-5-trifluoromethyl bromobenzene, 3-fluoro-5-trifluoromethylbromobenzene, 5-fluoro-3-bromobenzotrifluoride, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
1-bromo-3-fluoro-5-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents