Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzotrifluoride, 97%

$71.72 - $71.72

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-fluorobenzotrifluoride
CAS 130723-13-6
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00042498
Synonym 1-Bromo-3-fluoro-5-(trifluoromethyl)benzene; 3-Fluoro-5-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1803403

  SDS
alfa aesar™
A1803403
1g Each for $71.72
Description & Specifications

Specifications

Formula Weight 243
Assay 97%
CAS 130723-13-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 146°C
Synonym 1-Bromo-3-fluoro-5-(trifluoromethyl)benzene; 3-Fluoro-5-(trifluoromethyl)bromobenzene
Molecular Formula C7H3BrF4
Chemical Name or Material 3-Bromo-5-fluorobenzotrifluoride
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00042498
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.