Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzamide, 96%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-fluorobenzamide
CAS 933585-20-7
Formula Weight 218.02
MDL Number MFCD07780673
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6294403

 
alfa aesar™
H6294403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6294406

 
alfa aesar™
H6294406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62944MD

 
alfa aesar™
H62944MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.02
Physical Form Solid
CAS 933585-20-7
Assay 96%
Quantity 1g
MDL Number MFCD07780673
Chemical Name or Material 3-Bromo-5-fluorobenzamide