missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzaldehyde, 95%

Manufacturer:  Alfa Aesar™ H6404206

 View more versions of this product

Catalog No. AAH6404206



Description

Description

Applications
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air , heat.
Specifications

Specifications

3-Bromo-5-fluorobenzaldehyde
Light Sensitive
5g
C7H4BrFO
13904428
MEFQRXHVMJPOKZ-UHFFFAOYSA-N
3-bromo-5-fluorobenzaldehyde
21986246
Solid
1.43g/mL
41°C to 43°C
188813-02-7
MFCD04116319
3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde
C1=C(C=C(C=C1F)Br)C=O
203.01
203.01
95%

Chemical Identifiers

188813-02-7
203.01
MEFQRXHVMJPOKZ-UHFFFAOYSA-N
21986246
C1=C(C=C(C=C1F)Br)C=O
C7H4BrFO
MFCD04116319
3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde
3-bromo-5-fluorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents