Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluorobenzaldehyde, 95%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-fluorobenzaldehyde
CAS 188813-02-7
Formula Weight 203.01
MDL Number MFCD04116319
Beilstein 13904428
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6404206

  SDS
alfa aesar™
H6404206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6404214

  SDS
alfa aesar™
H6404214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6404222

  SDS
alfa aesar™
H6404222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 41° to 43°C
Formula Weight 203.01
Chemical Name or Material 3-Bromo-5-fluorobenzaldehyde
Density 1.43g/mL
Physical Form Solid
Assay 95%
Quantity 5g
Sensitivity Light Sensitive
MDL Number MFCD04116319
CAS 188813-02-7
Beilstein 13904428
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C

Applications
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air , heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C