missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-fluoro-2-methoxypyridine, 95%

$33.16 - $388.61

Chemical Identifiers

CAS 884494-81-9
Molecular Formula C6H5BrFNO
Molecular Weight (g/mol) 206.014
MDL Number MFCD04972397
InChI Key HSWCOAGQYSBFAK-UHFFFAOYSA-N
Synonym 3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
PubChem CID 40427006
IUPAC Name 3-bromo-5-fluoro-2-methoxypyridine
SMILES COC1=C(C=C(C=N1)F)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64915MD View Documents Alfa Aesar™
H64915MD
250mg Each for $33.16
 
AAH6491503 View Documents Alfa Aesar™
H6491503
1g Each for $97.78
 
AAH6491506 View Documents Alfa Aesar™
H6491506
5g Each for $388.61
 
Specifications

Chemical Identifiers

884494-81-9
206.014
HSWCOAGQYSBFAK-UHFFFAOYSA-N
40427006
COC1=C(C=C(C=N1)F)Br
C6H5BrFNO
MFCD04972397
3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
3-bromo-5-fluoro-2-methoxypyridine

Specifications

Light Sensitive
884494-81-9
MFCD04972397
3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
COC1=C(C=C(C=N1)F)Br
206.014
206.01
95%
250mg
C6H5BrFNO
15497859
HSWCOAGQYSBFAK-UHFFFAOYSA-N
3-bromo-5-fluoro-2-methoxypyridine
40427006
Liquid
3-Bromo-5-fluoro-2-methoxypyridine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C