missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-fluoro-2-cyanopyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451430010
Specifications
3-Bromo-5-fluoro-2-cyanopyridine | |
88.0°C to 91.0°C | |
950670-18-5 | |
C6H2BrFN2 | |
OLOSREGULYFXDD-UHFFFAOYSA-N | |
3-bromo-5-fluoropyridine-2-carbonitrile | |
46941477 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
3-aromo-5-fluoropicolinonitrile, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, 3-bromo-5-fluoropicolinonitrile, abbypharma ap-18-5887 | |
C1=C(C=NC(=C1Br)C#N)F | |
201 | |
201 |
Chemical Identifiers
950670-18-5 | |
201 | |
3-aromo-5-fluoropicolinonitrile, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, 3-bromo-5-fluoropicolinonitrile, abbypharma ap-18-5887 | |
3-bromo-5-fluoropyridine-2-carbonitrile |
C6H2BrFN2 | |
OLOSREGULYFXDD-UHFFFAOYSA-N | |
46941477 | |
C1=C(C=NC(=C1Br)C#N)F |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS Signal Word: Warning