Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-ethoxy-4-hydroxybenzaldehyde, 97%

$29.39 - $104.81

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-ethoxy-4-hydroxybenzaldehyde
CAS 3111-37-3
Molecular Formula C9H9BrO3
Formula Weight 245.08
MDL Number MFCD00173904
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1992503

 
alfa aesar™
A1992503
1g Each for $29.39

AAA1992506

 
alfa aesar™
A1992506
5g Each for $104.81
Description & Specifications

Specifications

Formula Weight 245.08
Molecular Formula C9H9BrO3
Melting Point 141° to 143°C
Chemical Name or Material 3-Bromo-5-ethoxy-4-hydroxybenzaldehyde
Assay 97%
MDL Number MFCD00173904
CAS 3111-37-3
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.