Learn More
3-Bromo-5-(chloromethyl)pyridine hydrochloride, 96%, Thermo Scientific™
$108.95 - $318.78
Chemical Identifiers
CAS | 120277-69-2 |
---|---|
Molecular Formula | C6H5BrClN |
Molecular Weight (g/mol) | 206.47 |
MDL Number | MFCD10697600 |
InChI Key | NLPHAZLCNNDGPS-UHFFFAOYSA-N |
Synonym | 3-bromo-5-chloromethyl pyridine, pyridine, 3-bromo-5-chloromethyl, 5-bromonicotinyl chloride, 3-bromo-5-chloromethyl-pyridine, pyridine, 3-bromo-5-chloromethyl-9ci |
PubChem CID | 11701259 |
IUPAC Name | 3-bromo-5-(chloromethyl)pyridine |
SMILES | ClCC1=CC(Br)=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66131MD
|
Thermo Scientific Chemicals
H66131MD |
250 mg |
Each for $108.95
|
|
|||||
AAH6613103
|
Thermo Scientific Chemicals
H6613103 |
1 g |
Each for $318.78
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
120277-69-2 | |
206.47 | |
NLPHAZLCNNDGPS-UHFFFAOYSA-N | |
11701259 | |
ClCC1=CC(Br)=CN=C1 |
C6H5BrClN | |
MFCD10697600 | |
3-bromo-5-chloromethyl pyridine, pyridine, 3-bromo-5-chloromethyl, 5-bromonicotinyl chloride, 3-bromo-5-chloromethyl-pyridine, pyridine, 3-bromo-5-chloromethyl-9ci | |
3-bromo-5-(chloromethyl)pyridine |
Specifications
120277-69-2 | |
C6H5BrClN | |
250 mg | |
1562735 | |
NLPHAZLCNNDGPS-UHFFFAOYSA-N | |
3-bromo-5-(chloromethyl)pyridine | |
11701259 | |
96% | |
3-Bromo-5-(chloromethyl)pyridine hydrochloride |
White | |
MFCD10697600 | |
UN3261 | |
3-bromo-5-chloromethyl pyridine, pyridine, 3-bromo-5-chloromethyl, 5-bromonicotinyl chloride, 3-bromo-5-chloromethyl-pyridine, pyridine, 3-bromo-5-chloromethyl-9ci | |
ClCC1=CC(Br)=CN=C1 | |
206.47 | |
242.93 | |
Solid |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures