missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-chlorobenzotrifluoride, 98%

$102.41 - $359.31

Chemical Identifiers

CAS 928783-85-1
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD09260894
InChI Key QRPRXRQFNNXPBA-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorobenzotrifluoride, 1-bromo-3-chloro-5-trifluoromethyl benzene, benzene, 1-bromo-3-chloro-5-trifluoromethyl, 5-bromo-3-chlorobenzotrifluoride, 3-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, acmc-209ri6, ksc912o0j, 3-bromo-5-chloro-trifluoromethylbenzene
PubChem CID 20269855
IUPAC Name 1-bromo-3-chloro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1Cl)Br)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6190614 View Documents Alfa Aesar™
H6190614
25g Each for $102.41
 
AAH6190622 View Documents Alfa Aesar™
H6190622
100g Each for $359.31
 
Specifications

Chemical Identifiers

928783-85-1
259.45
QRPRXRQFNNXPBA-UHFFFAOYSA-N
20269855
C1=C(C=C(C=C1Cl)Br)C(F)(F)F
C7H3BrClF3
MFCD09260894
3-bromo-5-chlorobenzotrifluoride, 1-bromo-3-chloro-5-trifluoromethyl benzene, benzene, 1-bromo-3-chloro-5-trifluoromethyl, 5-bromo-3-chlorobenzotrifluoride, 3-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, acmc-209ri6, ksc912o0j, 3-bromo-5-chloro-trifluoromethylbenzene
1-bromo-3-chloro-5-(trifluoromethyl)benzene

Specifications

25g
C7H3BrClF3
3-bromo-5-chlorobenzotrifluoride, 1-bromo-3-chloro-5-trifluoromethyl benzene, benzene, 1-bromo-3-chloro-5-trifluoromethyl, 5-bromo-3-chlorobenzotrifluoride, 3-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, acmc-209ri6, ksc912o0j, 3-bromo-5-chloro-trifluoromethylbenzene
C1=C(C=C(C=C1Cl)Br)C(F)(F)F
259.45
259.45
3-Bromo-5-chlorobenzotrifluoride
928783-85-1
MFCD09260894
QRPRXRQFNNXPBA-UHFFFAOYSA-N
1-bromo-3-chloro-5-(trifluoromethyl)benzene
20269855
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning