missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-chlorobenzonitrile 98.0+%, TCI America™
$66.37 - $204.06
Chemical Identifiers
CAS | 304854-55-5 |
---|---|
Molecular Formula | C7H3BrClN |
Molecular Weight (g/mol) | 216.462 |
MDL Number | MFCD06200840 |
InChI Key | DRKWKPSNVQVDKZ-UHFFFAOYSA-N |
PubChem CID | 15965893 |
IUPAC Name | 3-bromo-5-chlorobenzonitrile |
SMILES | C1=C(C=C(C=C1Cl)Br)C#N |
Chemical Identifiers
304854-55-5 | |
216.462 | |
DRKWKPSNVQVDKZ-UHFFFAOYSA-N | |
3-bromo-5-chlorobenzonitrile |
C7H3BrClN | |
MFCD06200840 | |
15965893 | |
C1=C(C=C(C=C1Cl)Br)C#N |
Specifications
304854-55-5 | |
White-Yellow | |
C7H3BrClN | |
1 g | |
DRKWKPSNVQVDKZ-UHFFFAOYSA-N | |
3-bromo-5-chlorobenzonitrile | |
15965893 | |
≥98.0% (GC) | |
3-Bromo-5-chlorobenzonitrile |
74°C | |
144°C | |
MFCD06200840 | |
3439 | |
C1=C(C=C(C=C1Cl)Br)C#N | |
216.462 | |
216.46 | |
Crystalline Powder |
Safety and Handling
TSCA : No