Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-chloro-2,6-dimethoxybenzamide, 97%

$124.00 - $436.96

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-chloro-2,6-dimethoxybenzamide
CAS 175135-60-1
Molecular Formula C9H9BrClNO3
Formula Weight 294.53
MDL Number MFCD00052965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2473503

  SDS
alfa aesar™
B2473503
1g Each for $124.00

AAB2473506

  SDS
alfa aesar™
B2473506
5g Each for $436.96
Description & Specifications

Specifications

Formula Weight 294.53
Quantity 1g
MDL Number MFCD00052965
Molecular Formula C9H9BrClNO3
Chemical Name or Material 3-Bromo-5-chloro-2,6-dimethoxybenzamide
Assay 97%
CAS 175135-60-1