Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid
Formula Weight 311.97
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6111303

 
alfa aesar™
H6111303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61113MD

 
alfa aesar™
H61113MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 311.97
Chemical Name or Material 3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid
Quantity 1g
Physical Form Solid
Assay 97%