Please login to your online account to display your discounted pricing

3-Bromo-4-pyridinecarboxaldehyde, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-pyridinecarboxaldehyde
Assay Percent Range 97%
Molecular Formula C6H4BrNO
Formula Weight 186
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398080010

  SDS
thermo fisher scientific
AC398080010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398080050

  SDS
thermo fisher scientific
AC398080050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186
Quantity 1g
Chemical Name or Material 3-Bromo-4-pyridinecarboxaldehyde
CAS 70201-43-3
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H4BrNO