Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-nitropyridine N-oxide, 98+%

$176.30 - $656.95

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-nitropyridine N-oxide
CAS 1678-49-5
Molecular Formula C5H3BrN2O3
Formula Weight 219
MDL Number MFCD00085925
Beilstein 150822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1952406

  SDS
alfa aesar™
L1952406
5g Each for $176.30

AAL1952414

  SDS
alfa aesar™
L1952414
25g Each for $656.95
Description & Specifications

Specifications

Formula Weight 219
Chemical Name or Material 3-Bromo-4-nitropyridine N-oxide
Assay ≥98%
Molecular Formula C5H3BrN2O3
MDL Number MFCD00085925
Quantity 5g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1678-49-5
Melting Point 152° to 154°C
Beilstein 150822
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Sparingly Soluble in water (0.92 g/L at 25°C).

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.