missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-nitrobenzotrifluoride, 95%

$30.99 - $362.39

Chemical Identifiers

CAS 132839-58-8
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.005
MDL Number MFCD09998160
InChI Key DJOLBPUSSIOMKU-UHFFFAOYSA-N
Synonym 2-bromo-1-nitro-4-trifluoromethyl benzene, 3-bromo-4-nitrobenzotrifluoride, 2-bromo-4-trifluoromethyl nitrobenzene, benzene, 2-bromo-1-nitro-4-trifluoromethyl, acmc-209bq5, ksc496a5t, 2-bromo-1-nitro-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethylnitrobenzene
PubChem CID 14790878
IUPAC Name 2-bromo-1-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64376MD View Documents Alfa Aesar™
H64376MD
250mg Each for $30.99
 
AAH6437603 View Documents Alfa Aesar™
H6437603
1g Each for $91.30
 
AAH6437606 View Documents Alfa Aesar™
H6437606
5g Each for $362.39
 
Specifications

Chemical Identifiers

132839-58-8
270.005
DJOLBPUSSIOMKU-UHFFFAOYSA-N
14790878
C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]
C7H3BrF3NO2
MFCD09998160
2-bromo-1-nitro-4-trifluoromethyl benzene, 3-bromo-4-nitrobenzotrifluoride, 2-bromo-4-trifluoromethyl nitrobenzene, benzene, 2-bromo-1-nitro-4-trifluoromethyl, acmc-209bq5, ksc496a5t, 2-bromo-1-nitro-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethylnitrobenzene
2-bromo-1-nitro-4-(trifluoromethyl)benzene

Specifications

Yellow
132839-58-8
MFCD09998160
DJOLBPUSSIOMKU-UHFFFAOYSA-N
2-bromo-1-nitro-4-(trifluoromethyl)benzene
14790878
Liquid
3-Bromo-4-nitrobenzotrifluoride
250mg
C7H3BrF3NO2
2-bromo-1-nitro-4-trifluoromethyl benzene, 3-bromo-4-nitrobenzotrifluoride, 2-bromo-4-trifluoromethyl nitrobenzene, benzene, 2-bromo-1-nitro-4-trifluoromethyl, acmc-209bq5, ksc496a5t, 2-bromo-1-nitro-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethylnitrobenzene
C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]
270.005
270.01
95%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning