missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2726806

 View more versions of this product

Catalog No. AAH2726806



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-4-nitrobenzoic acid
200°C to 204°C
MFCD04117949
3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
246.02
246.02
101420-81-9
C7H4BrNO4
5 g
KKPPNEJUUOQRLE-UHFFFAOYSA-N
3-bromo-4-nitrobenzoic acid
16218087
97%

Chemical Identifiers

101420-81-9
246.02
KKPPNEJUUOQRLE-UHFFFAOYSA-N
16218087
OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
C7H4BrNO4
MFCD04117949
3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
3-bromo-4-nitrobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only