Please login to your online account to display your discounted pricing

3-Bromo-4-methylbenzotrifluoride, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC446680250

 View more versions of this product

Catalog No. AC446680250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 239.03
Quantity 25g
Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
Assay Percent Range 97%
Molecular Formula C8H6BrF3
CAS 66417-30-9
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

Specifications

Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
Assay Percent Range 97%
Molecular Formula C8H6BrF3
Formula Weight 239.03
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.