missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-methylbenzotrifluoride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446680050

 View more versions of this product

Catalog No. AC446680050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Bromo-4-methylbenzotrifluoride
5g
96% min. (GC)
2-bromo-1-methyl-4-trifluoromethyl benzene, 2-bromo-1-methyl-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethyl toluene, 2-bromo-4-trifluoromethyltoluene, 2-methyl-5-trifluoromethyl bromobenzene, 3-bromo-4-methylbenzotrifluoride, acmc-209nvg, benzene, 2-bromo-1-methyl-4-trifluoromethyl, ksc294o7j
CC1=C(C=C(C=C1)C(F)(F)F)Br
239.03
239.03
88.0°C (31.0 Torr)
66417-30-9
C8H6BrF3
TWWUWLMGYSLODU-UHFFFAOYSA-N
2-bromo-1-methyl-4-(trifluoromethyl)benzene
2779376
97%

Chemical Identifiers

66417-30-9
239.03
2-bromo-1-methyl-4-trifluoromethyl benzene, 2-bromo-1-methyl-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethyl toluene, 2-bromo-4-trifluoromethyltoluene, 2-methyl-5-trifluoromethyl bromobenzene, 3-bromo-4-methylbenzotrifluoride, acmc-209nvg, benzene, 2-bromo-1-methyl-4-trifluoromethyl, ksc294o7j
2-bromo-1-methyl-4-(trifluoromethyl)benzene
C8H6BrF3
TWWUWLMGYSLODU-UHFFFAOYSA-N
2779376
CC1=C(C=C(C=C1)C(F)(F)F)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

Documents