Please login to your online account to display your discounted pricing

3-Bromo-4-methylbenzotrifluoride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
Assay Percent Range 97%
Molecular Formula C8H6BrF3
Formula Weight 239.03
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC446680050

 
thermo fisher scientific
AC446680050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.
Health Hazard 2 GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Formula Weight 239.03
Quantity 5g
Health Hazard 1 Warning
Molecular Formula C8H6BrF3
CAS 66417-30-9
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride