Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-methylbenzotrifluoride, 97%

$138.54 - $772.72

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
CAS 66417-30-9
Molecular Formula C8H6BrF3
Formula Weight 239.04
MDL Number MFCD01631584
Synonym 2-Bromo-4-(trifluoromethyl)toluene; 2-Methyl-5-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2632803

  SDS
alfa aesar™
H2632803
1g Each for $138.54

AAH2632809

  SDS
alfa aesar™
H2632809
10g Each for $772.72
Description & Specifications

Specifications

Formula Weight 239.04
Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
Assay 97%
Molecular Formula C8H6BrF3
MDL Number MFCD01631584
CAS 66417-30-9
Synonym 2-Bromo-4-(trifluoromethyl)toluene; 2-Methyl-5-(trifluoromethyl)bromobenzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.