Learn More
3-Bromo-4-methylbenzonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2780614
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
3-Bromo-4-methylbenzonitrile | |
39°C to 43°C | |
C8H6BrN | |
25 g | |
2-bromo-4-cyanotoluene, 4-methyl-3-bromobenzonitrile, benzonitrile, 3-bromo-4-methyl, 3-bromo-4-methylbenzenecarbonitrile, 3-bromo-4-methyl-benzonitrile, bromo p tolunitrile, bromo-p-tolunitrile, bromo-p-toluonitrile, 3-bromo-p-tolunitrile, pubchem11163 | |
VXUMRYMTYKDWMO-UHFFFAOYSA-N | |
3-bromo-4-methylbenzonitrile | |
14004840 | |
97% |
42872-74-2 | |
>110°C (230°F) | |
MFCD06797818 | |
UN3439 | |
Insoluble in water. | |
CC1=C(Br)C=C(C=C1)C#N | |
196.05 | |
196.05 |
Chemical Identifiers
42872-74-2 | |
196.05 | |
VXUMRYMTYKDWMO-UHFFFAOYSA-N | |
14004840 | |
CC1=C(Br)C=C(C=C1)C#N |
C8H6BrN | |
MFCD06797818 | |
2-bromo-4-cyanotoluene, 4-methyl-3-bromobenzonitrile, benzonitrile, 3-bromo-4-methyl, 3-bromo-4-methylbenzenecarbonitrile, 3-bromo-4-methyl-benzonitrile, bromo p tolunitrile, bromo-p-tolunitrile, bromo-p-toluonitrile, 3-bromo-p-tolunitrile, pubchem11163 | |
3-bromo-4-methylbenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only