missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-methylbenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1149622

 View more versions of this product

Catalog No. AAA1149622



Description

Description

Applications
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Specifications

3-Bromo-4-methylbenzoic acid
206°C to 211°C
7697-26-9
MFCD00002488
3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
CC1=C(C=C(C=C1)C(=O)O)Br
215.046
215.05
Odorless
100g
C8H7BrO2
1936510
ZFJOMUKPDWNRFI-UHFFFAOYSA-N
3-bromo-4-methylbenzoic acid
82130
≥98%

Chemical Identifiers

7697-26-9
215.046
ZFJOMUKPDWNRFI-UHFFFAOYSA-N
82130
CC1=C(C=C(C=C1)C(=O)O)Br
C8H7BrO2
MFCD00002488
3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
3-bromo-4-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

SDS Documents