missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-methylbenzaldehyde, TCI America™

$44.31 - $149.72

Chemical Identifiers

CAS 36276-24-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD04971197
InChI Key WTXXUAHMTVAQHW-UHFFFAOYSA-N
PubChem CID 816765
IUPAC Name 3-bromo-4-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B50731G View Documents TCI America
B50731G
1g Each for $44.31
 
B50735G View Documents TCI America
B50735G
5g Each for $149.72
 
Specifications

Chemical Identifiers

36276-24-1
199.047
WTXXUAHMTVAQHW-UHFFFAOYSA-N
3-bromo-4-methylbenzaldehyde
C8H7BrO
MFCD04971197
816765
CC1=C(C=C(C=C1)C=O)Br

Specifications

White
1g
C8H7BrO
WTXXUAHMTVAQHW-UHFFFAOYSA-N
3-bromo-4-methylbenzaldehyde
816765
Crystal-Powder at 20°C
3-Bromo-4-methylbenzaldehyde
48°C
36276-24-1
MFCD04971197
CC1=C(C=C(C=C1)C=O)Br
199.047
199.05
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No