Learn More
3-Bromo-4-methoxypyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H64296MD
Description
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water (9.8 g/L).
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
3-Bromo-4-methoxypyridine | |
Yellow | |
MFCD05664024 | |
1423610 | |
Slightly soluble in water (9.8g/L). | |
COC1=C(C=NC=C1)Br | |
188.024 | |
188.02 | |
Liquid |
82257-09-8 | |
C6H6BrNO | |
250 mg | |
3-bromo-4-methoxy-pyridine, pyridine, 3-bromo-4-methoxy, pyridine,3-bromo-4-methoxy, pubchem6686, 4-methoxy-3-bromopyridine, acmc-209po0, abbypharma ap-17-5958 | |
DSFJKFHIMBVWAV-UHFFFAOYSA-N | |
3-bromo-4-methoxypyridine | |
642827 | |
97% |
Chemical Identifiers
82257-09-8 | |
188.024 | |
DSFJKFHIMBVWAV-UHFFFAOYSA-N | |
3-bromo-4-methoxypyridine |
C6H6BrNO | |
MFCD05664024 | |
642827 | |
COC1=C(C=NC=C1)Br |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only