Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-methoxybiphenyl, 99%

$99.14 - $340.53

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-methoxybiphenyl
CAS 74447-73-7
Molecular Formula C13H11BrO
Formula Weight 263.14
MDL Number MFCD00079720
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5058103

  SDS
alfa aesar™
H5058103
1g Each for $340.53

AAH50581MD

  SDS
alfa aesar™
H50581MD
250mg Each for $99.14
Description & Specifications

Specifications

Formula Weight 263.14
Chemical Name or Material 3-Bromo-4-methoxybiphenyl
Quantity 1g
Assay 99%
Molecular Formula C13H11BrO
MDL Number MFCD00079720
CAS 74447-73-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.