missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid, 95%

$73.45 - $293.20

Chemical Identifiers

CAS 1215206-32-8
Molecular Formula C14H11BrO3
Formula Weight 307.14
MDL Number MFCD15143539
Chemical Name or Material 3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5191203 SDS Alfa Aesar™
H5191203
1g Each for $73.45
AAH5191206 SDS Alfa Aesar™
H5191206
5g Each for $293.20
Specifications Description & Specifications

Chemical Identifiers

1215206-32-8
307.14
3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid
C14H11BrO3
MFCD15143539

Specifications

1215206-32-8
307.14
1g
95%
C14H11BrO3
MFCD15143539
3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning