missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-methoxybenzyl Cyanide, TCI America™

$142.09 - $443.06

Chemical Identifiers

CAS 772-59-8
Molecular Formula C9H8BrNO
Molecular Weight (g/mol) 226.073
MDL Number MFCD00016391
InChI Key OBJKHHRZMIIEOK-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxyphenylacetonitrile, 3-bromo-4-methoxybenzyl cyanide, 2-3-bromo-4-methoxyphenyl acetonitrile, benzeneacetonitrile, 3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethanenitrile, acmc-209p8t, benzeneacetonitrile,3-bromo-4-methoxy, 3-bromo-4-methoxy-phenyl-acetonitrile, 2-3-bromanyl-4-methoxy-phenyl ethanenitrile
PubChem CID 522655
IUPAC Name 2-(3-bromo-4-methoxyphenyl)acetonitrile
SMILES COC1=C(C=C(C=C1)CC#N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14555G View Documents TCI America
B14555G
5g Each for $142.09
 
B145525G View Documents TCI America
B145525G
25g Each for $443.06
 
Specifications

Chemical Identifiers

772-59-8
226.073
OBJKHHRZMIIEOK-UHFFFAOYSA-N
522655
COC1=C(C=C(C=C1)CC#N)Br
C9H8BrNO
MFCD00016391
3-bromo-4-methoxyphenylacetonitrile, 3-bromo-4-methoxybenzyl cyanide, 2-3-bromo-4-methoxyphenyl acetonitrile, benzeneacetonitrile, 3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethanenitrile, acmc-209p8t, benzeneacetonitrile,3-bromo-4-methoxy, 3-bromo-4-methoxy-phenyl-acetonitrile, 2-3-bromanyl-4-methoxy-phenyl ethanenitrile
2-(3-bromo-4-methoxyphenyl)acetonitrile

Specifications

178°C
56°C
772-59-8
MFCD00016391
3-bromo-4-methoxyphenylacetonitrile, 3-bromo-4-methoxybenzyl cyanide, 2-3-bromo-4-methoxyphenyl acetonitrile, benzeneacetonitrile, 3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethanenitrile, acmc-209p8t, benzeneacetonitrile,3-bromo-4-methoxy, 3-bromo-4-methoxy-phenyl-acetonitrile, 2-3-bromanyl-4-methoxy-phenyl ethanenitrile
COC1=C(C=C(C=C1)CC#N)Br
226.073
226.07
≥98.0% (GC)
Yellow
5g
C9H8BrNO
2811
OBJKHHRZMIIEOK-UHFFFAOYSA-N
2-(3-bromo-4-methoxyphenyl)acetonitrile
522655
Crystal-Powder at 20°C
3-Bromo-4-methoxybenzyl Cyanide
Safety and Handling

Safety and Handling

TSCA : No