Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-methoxybenzonitrile, 99%

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-methoxybenzonitrile
CAS 117572-79-9
Molecular Formula C8H6BrNO
Formula Weight 212.05
MDL Number MFCD00143097
UN Number UN3439
Beilstein 3246576
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1669806

 
alfa aesar™
A1669806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1669814

 
alfa aesar™
A1669814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1669822

 
alfa aesar™
A1669822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.05
Chemical Name or Material 3-Bromo-4-methoxybenzonitrile
Assay 99%
Molecular Formula C8H6BrNO
MDL Number MFCD00143097
CAS 117572-79-9
UN Number UN3439
Melting Point 120° to 124°C
Beilstein 3246576
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
3-Bromo-4-methoxybenzonitrile is used to prepare 3-bromo-2-iodo-4-methoxy-benzonitrile. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.