missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-methoxybenzenesulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H54015MD

 View more versions of this product

Catalog No. AAH54015MD



Specifications

Specifications

3-Bromo-4-methoxybenzenesulfonyl chloride
250mg
C7H6BrClO3S
UN3265
LTVLATJRJIBHDR-UHFFFAOYSA-N
3-bromo-4-methoxybenzenesulfonyl chloride
3863747
97%
Moisture sensitive
23094-96-4
MFCD03989643
3-bromo-4-methoxy-benzene-sulfonyl chloride
COC1=C(C=C(C=C1)S(=O)(=O)Cl)Br
285.536
285.54

Chemical Identifiers

23094-96-4
285.536
LTVLATJRJIBHDR-UHFFFAOYSA-N
3863747
COC1=C(C=C(C=C1)S(=O)(=O)Cl)Br
C7H6BrClO3S
MFCD03989643
3-bromo-4-methoxy-benzene-sulfonyl chloride
3-bromo-4-methoxybenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents