missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-isocyanatothiophene, 95%, Maybridge

Chemical Identifiers

CAS 937795-88-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C5H2BrNOS
Formula Weight 204.05
MDL Number MFCD09879904
Chemical Name or Material 3-Bromo-4-isocyanatothiophene
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC54206DA SDS Fisher Scientific Company
CC54206DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC54206DE SDS Fisher Scientific Company
CC54206DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

937795-88-5
100.0
C5H2BrNOS
MFCD09879904
95%
94.5
95%
204.05
3-Bromo-4-isocyanatothiophene

Specifications

937795-88-5
100.0
1g
C5H2BrNOS
MFCD09879904
95%
94.5
Amber Glass Bottle
95%
204.05
3-Bromo-4-isocyanatothiophene

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger