Please login to your online account to display your discounted pricing

3-Bromo-4-hydroxy-5-methoxybenzaldehyde 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB06293EA

 View more versions of this product

Catalog No. BTB06293EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.04
Quantity 10g
Assay 97%
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C8H7BrO3
Linear Formula BrC6H2(OH)(OCH3)CHO
CAS 2973-76-4

Specifications

Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7BrO3
Linear Formula BrC6H2(OH)(OCH3)CHO
Formula Weight 231.04