missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-hydroxy-5-methoxybenzaldehyde 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C8H7BrO3
Linear Formula BrC6H2(OH)(OCH3)CHO
Formula Weight 231.04
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB06293EA Fisher Scientific Company
BTB06293EA
Amber glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
BrC6H2(OH)(OCH3)CHO
3-Bromo-4-hydroxy-5-methoxybenzaldehyde
C8H7BrO3
231.04
97%

Specifications

231.04
BrC6H2(OH)(OCH3)CHO
Amber glass bottle
2973-76-4
97%
10g
97%
C8H7BrO3
3-Bromo-4-hydroxy-5-methoxybenzaldehyde