Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-fluorotoluene, 99%

$47.91 - $197.39

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-fluorotoluene
CAS 452-62-0
Molecular Formula C7H6BrF
Formula Weight 189.03
MDL Number MFCD00040827
Beilstein 1680604
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2152306

  SDS
alfa aesar™
B2152306
5g Each for $47.91

AAB2152314

  SDS
alfa aesar™
B2152314
25g Each for $197.39
Description & Specifications

Specifications

Formula Weight 189.03
Chemical Name or Material 3-Bromo-4-fluorotoluene
Assay 99%
Boiling Point 168° to 170°C
Quantity 5g
Flash Point 73°C (163°F)
Molecular Formula C7H6BrF
MDL Number MFCD00040827
CAS 452-62-0
Beilstein 1680604
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.