Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Bromo-4'-fluoropropiophenone, 98%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 3'-Bromo-4'-fluoropropiophenone
CAS 202865-82-5
Molecular Formula C9H8BrFO
Formula Weight 231.06
MDL Number MFCD00143012
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2542914

 
alfa aesar™
B2542914
25g Each for $213.96
Description & Specifications

Specifications

Formula Weight 231.06
Chemical Name or Material 3'-Bromo-4'-fluoropropiophenone
Assay 98%
Molecular Formula C9H8BrFO
MDL Number MFCD00143012
CAS 202865-82-5
Melting Point 60° to 64°C
Quantity 25g