missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-fluorophenylacetic Acid, TCI America™

$96.25 - $342.23

Chemical Identifiers

CAS 194019-11-9
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD04038373
InChI Key XXFGIJYSXNXNAU-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid
PubChem CID 3416339
IUPAC Name 2-(3-bromo-4-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29765G View Documents TCI America
B29765G
5g Each for $96.25
 
B297625G View Documents TCI America
B297625G
25g Each for $342.23
 
Specifications

Chemical Identifiers

194019-11-9
233.036
XXFGIJYSXNXNAU-UHFFFAOYSA-N
3416339
C1=CC(=C(C=C1CC(=O)O)Br)F
C8H6BrFO2
MFCD04038373
3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid
2-(3-bromo-4-fluorophenyl)acetic acid

Specifications

White-Yellow
5g
C8H6BrFO2
3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid
C1=CC(=C(C=C1CC(=O)O)Br)F
233.036
233.04
≥97.0% (GC,T)
67°C
194019-11-9
MFCD04038373
XXFGIJYSXNXNAU-UHFFFAOYSA-N
2-(3-bromo-4-fluorophenyl)acetic acid
3416339
Crystal-Powder at 20°C
3-Bromo-4-fluorophenylacetic Acid
Safety and Handling

Safety and Handling

TSCA : No