Please login to your online account to display your discounted pricing

3-Bromo-4-fluorocinnamic acid, 99+%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC290540050

 View more versions of this product

Catalog No. AC290540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Chunks
Formula Weight 245.05
Quantity 5g
Color Light yellow
TSCA TSCA
Melting Point 189° to 192°C
CAS Min % 99.0
CAS Max % 100.0
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C9 H6 Br F O2
Chemical Name or Material 3-Bromo-4-fluorocinnamic acid
Packaging Glass bottle
MDL Number MFCD00143095
CAS 160434-49-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Bromo-4-fluorocinnamic acid
Name Note 99+%
CAS 160434-49-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9 H6 Br F O2
Formula Weight 245.05
MDL Number MFCD00143095
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents