missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-fluorocinnamic acid, 99+%, ACROS Organics™

Chemical Identifiers

CAS 160434-49-1
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C9 H6 Br F O2
Formula Weight 245.05
MDL Number MFCD00143095
Chemical Name or Material 3-Bromo-4-fluorocinnamic acid, 99+%
Assay ≥99.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC290540050 SDS Thermo Fisher Scientific
AC290540050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

160434-49-1
100.0
C9 H6 Br F O2
MFCD00143095
≥99.0%
99.0
99+%
245.05
3-Bromo-4-fluorocinnamic acid, 99+%

Specifications

160434-49-1
100.0
Glass bottle
189° to 192°C
99+%
C9 H6 Br F O2
MFCD00143095
≥99.0%
99.0
Authentic
Light yellow
Crystalline Chunks
5g
245.05
3-Bromo-4-fluorocinnamic acid, 99+%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA