Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-fluorocinnamic acid, 98%

$54.55 - $735.30

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-fluorocinnamic acid
CAS 160434-49-1
Molecular Formula C9H6BrFO2
Formula Weight 245.05
MDL Number MFCD00143095
Beilstein 8977970
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1452606

  SDS
alfa aesar™
A1452606
Each for $54.55

AAA1452614

  SDS
alfa aesar™
A1452614
Each for $219.99

AAA1452622

  SDS
alfa aesar™
A1452622
Each for $735.30
Description & Specifications

Specifications

Formula Weight 245.05
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 3-Bromo-4-fluorocinnamic acid
Assay 98%
Melting Point 193° to 195°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 160434-49-1
MDL Number MFCD00143095
Molecular Formula C9H6BrFO2
Quantity 5g
Beilstein 8977970
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.