missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-fluorocinnamic acid, 98%

Manufacturer:  Alfa Aesar™ A1452614

 View more versions of this product

Catalog No. AAA1452614



Specifications

Specifications

3-Bromo-4-fluorocinnamic acid
25g
C9H6BrFO2
8977970
ZNIGVADAKXOMQH-DUXPYHPUSA-N
(E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid
688312
98%
193°C to 195°C
160434-49-1
MFCD00143095
3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391
C1=CC(=C(C=C1C=CC(=O)O)Br)F
245.047
245.05

Chemical Identifiers

160434-49-1
245.047
ZNIGVADAKXOMQH-DUXPYHPUSA-N
688312
C1=CC(=C(C=C1C=CC(=O)O)Br)F
C9H6BrFO2
MFCD00143095
3-bromo-4-fluorocinnamic acid, 2e-3-3-bromo-4-fluorophenyl prop-2-enoic acid, e-3-3-bromo-4-fluorophenyl acrylic acid, 3-3-bromo-4-fluoro-phenyl-acrylic acid, 3-3-bromo-4-fluorophenyl acrylic acid, pubchem3228, 2e-3-3-bromo-4-fluorophenyl acrylic acid, rarechem bk hw 0193, timtec-bb sbb003187, attercop-chm at116391
(E)-3-(3-bromo-4-fluorophenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents