Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-fluorobiphenyl, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-fluorobiphenyl
CAS 306935-88-6
Molecular Formula C12H8BrF
Formula Weight 251.1
MDL Number MFCD01571091
UN Number UN3151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2531606

 
alfa aesar™
B2531606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2531614

 
alfa aesar™
B2531614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.1
Chemical Name or Material 3-Bromo-4-fluorobiphenyl
Assay 98%
Quantity 5g
Molecular Formula C12H8BrF
MDL Number MFCD01571091
CAS 306935-88-6
UN Number UN3151
DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID